3-(2-(((4-Carbamimidoylphenyl)amino)methyl)-1-methyl-N-(pyridin-2-yl)-1H-benzo[d]imidazole-5-carboxamido)propanoic acid 98%

Supplier: Ambeed
Warning

A112634-100MG A112634-10MG A112634-1G A112634-250MG A112634-2MG A112634-50MG A112634-5G A112634-5MG
76781-196EA 40.32 USD
76781-196 76781-198 76781-200 76781-202 76781-204 76781-206 76781-208 76781-210
3-(2-(((4-Carbamimidoylphenyl)amino)methyl)-1-methyl-N-(pyridin-2-yl)-1H-benzo[d]imidazole-5-carboxamido)propanoic acid 98%
3-(2-(((4-Carbamimidoylphenyl)amino)methyl)-1-methyl-N-(pyridin-2-yl)-1H-benzo[d]imidazole-5-carboxamido)propanoic acid

Precaución: for Research Use Only

Formula: C₂₅H₂₅N₇O₃
MW: 471.51 g/mol
MDL Number: MFCD09837830
CAS Number: 211914-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR