trans,trans-4-(3,4-Difluorophenyl)-4'-ethyl-1,1'-bi(cyclohexane) 97%

Supplier: Ambeed
Warning

A112590-100G A112590-10G A112590-25G A112590-500G A112590-5G
76781-186EA 82.76 USD
76781-186 76781-188 76781-190 76781-192 76781-194
trans,trans-4-(3,4-Difluorophenyl)-4'-ethyl-1,1'-bi(cyclohexane) 97%
trans,trans-4-(3,4-Difluorophenyl)-4'-ethyl-1,1'-bi(cyclohexane)

Precaución: for Research Use Only

Formula: C₂₀H₂₈F₂
MW: 306.43 g/mol
MDL Number: MFCD13188635
CAS Number: 118164-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR