Terephthalonitrile 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
1,4-Benzenedicarbonitrile, Terephthalodinitrile, Benzene-1,4-dicarbonitrile, 1,4-Dicyanobenzene
A112242-1000G
A112242-100G
A112242-25G
A112242-500G
76781-166EA
114.58
USD
76781-166
76781-168
76781-170
76781-172
Terephthalonitrile 95%
Terephthalonitrile
Precaución: for Research Use Only
Formula:
C₈H₄N₂ MW: 128.13 g/mol |
MDL Number:
MFCD00001810 CAS Number: 623-26-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...