N-Boc-L-phenylalaninal 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (S)-(1-oxo-3-phenylpropan-2-yl)carbamate

A111795-1G A111795-5G
76781-146EA 52 USD
76781-146 76781-148
N-Boc-L-phenylalaninal 95%
N-Boc-L-phenylalaninal

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO₃
MW: 249.31 g/mol
MDL Number: MFCD00143854
CAS Number: 72155-45-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR