EEDQ (ethyl-N-(2-ethoxy-1,2-dihydroquinoline)carboxylate) 95%

Supplier: Ambeed
Warning

Synonyms: 2-Ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline, N-Ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, Ethyl 2-ethoxyquinoline-1(2H)-carboxylate

A111741-100G A111741-10G A111741-25G A111741-500G
76781-138EA 34 USD
76781-138 76781-140 76781-142 76781-144
EEDQ (ethyl-N-(2-ethoxy-1,2-dihydroquinoline)carboxylate) 95%
EEDQ (ethyl-N-(2-ethoxy-1,2-dihydroquinoline)carboxylate)

Precaución: for Research Use Only

Formula: C₁₄H₁₇NO₃
MW: 247.29 g/mol
MDL Number: MFCD00006703
CAS Number: 16357-59-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR