(1-Ethoxycarbonylethyl)triphenylphosphonium bromide 97%

Supplier: Ambeed
Warning

Synonyms: (1-Ethoxy-1-oxopropan-2-yl)triphenylphosphonium bromide

A111740-100G A111740-10G A111740-25G A111740-500G
76781-130EA 51 USD
76781-130 76781-132 76781-134 76781-136
(1-Ethoxycarbonylethyl)triphenylphosphonium bromide 97%
(1-Ethoxycarbonylethyl)triphenylphosphonium bromide

Precaución: for Research Use Only

Formula: C₂₃H₂₄BrO₂P
MW: 443.33 g/mol
MDL Number: MFCD00031605
CAS Number: 30018-16-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR