trans-N-(6,6-Dimethyl-2-hepten-4-ynyl)-N-methyl-1-naphthylmethylamine hydrochloride 99%

Supplier: Ambeed
Warning

Synonyms: Terbinafine Hydrochloride, Terbinafine HCl, (E)-N,6,6-Trimethyl-N-(naphthalen-1-ylmethyl)hept-2-en-4-yn-1-amine hydrochloride

A111542-100G A111542-10G A111542-1MG A111542-25G A111542-5G
76781-118EA 90 USD
76781-118 76781-120 76781-122 76781-124 76781-126
trans-N-(6,6-Dimethyl-2-hepten-4-ynyl)-N-methyl-1-naphthylmethylamine hydrochloride 99%
trans-N-(6,6-Dimethyl-2-hepten-4-ynyl)-N-methyl-1-naphthylmethylamine hydrochloride

Precaución: for Research Use Only

Formula: C₂₁H₂₅N
MDL Number: MFCD00145430
CAS Number: 78628-80-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR