5-Amino-4-chloro-6-methoxypyrimidine 97%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-6-methoxypyrimidin-5-amine

A111445-1G A111445-250MG A111445-5G
76781-112EA 57.29 USD
76781-112 76781-114 76781-116
5-Amino-4-chloro-6-methoxypyrimidine 97%
5-Amino-4-chloro-6-methoxypyrimidine

Precaución: for Research Use Only

Formula: C₅H₆ClN₃O
MW: 159.574 g/mol
MDL Number: MFCD01646119
CAS Number: 15846-19-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR