Ethyl-2-(3-bromophenyl)acetate 97%

Supplier: Ambeed
Warning

A111400-100G A111400-1G A111400-25G A111400-5G A111400-10G
76781-106EA 131 USD
76781-106 76781-108 76781-110 77666-626 77871-453
Ethyl-2-(3-bromophenyl)acetate 97%
Ethyl-2-(3-bromophenyl)acetate

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₂
MDL Number: MFCD09953130
CAS Number: 14062-30-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR