Tetrabutylammonium tribromide 97%

Supplier: Ambeed
Danger

Synonyms: Tetra-n-butylammonium tribromide, N,N,N,N-Tetrabutylammonium tribromide

A111134-10G A111134-25G A111134-100G
76781-092EA 74.27 USD
76781-092 76781-094 76781-096 77599-600
Tetrabutylammonium tribromide 97%
Tetrabutylammonium tribromide

Precaución: for Research Use Only

Formula: C₁₆H₃₆Br₃N
MW: 482.18 g/mol
Melting Pt: 76
MDL Number: MFCD00012110
CAS Number: 38932-80-8
UN: 1759
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR