Methyl 2-chloro-5-hydroxybenzoate 97%

Supplier: Ambeed
Warning

A111023-1G A111023-250MG A111023-5G
76781-080EA 110.34 USD
76781-080 76781-082 76781-084
Methyl 2-chloro-5-hydroxybenzoate 97%
Methyl 2-chloro-5-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₈H₇ClO₃
MW: 186.592 g/mol
MDL Number: MFCD10698699
CAS Number: 247092-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR