2-Chloro-4-Bromo-5-fluorobenzaldehyde 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Chloro-4-bromo-5-fluorobenzaldehyde, 4-Bromo-2-chloro-5-fluorobenzaldehyde
A111007-1G
A111007-5G
76781-078EA
44
USD
76781-078
77570-628
2-Chloro-4-Bromo-5-fluorobenzaldehyde 95%
2-Chloro-4-Bromo-5-fluorobenzaldehyde
Precaución: for Research Use Only
Formula:
C₇H₃BrClFO MW: 237.452 g/mol |
MDL Number:
MFCD13185877 CAS Number: 1214386-29-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...