3-Cyano-4-fluorophenylboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 3-Cyano-4-fluorobenzeneboronic acid pinacol ester, 2-(3-Cyano-4-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Cyano-4-fluorophenylboronic acid Pinacol ester, 3-Cyano-4-fluorophenylboronic acid, pinacol ester, 2-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile

A110977-100G A110977-10G A110977-1G A110977-250MG A110977-25G A110977-5G
76781-066EA 407 USD
76781-066 76781-068 76781-070 76781-072 76781-074 76781-076
3-Cyano-4-fluorophenylboronic acid pinacol ester 97%
3-Cyano-4-fluorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₅BFNO₂
MDL Number: MFCD06795681
CAS Number: 775351-57-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR