6-Methylpyrimidin-4-amine 97%

Supplier: Ambeed
Warning

A110074-100MG A110074-1G A110074-250MG A110074-5G
76781-038EA 24.41 USD
76781-038 76781-040 76781-042 76781-044
6-Methylpyrimidin-4-amine 97%
6-Methylpyrimidin-4-amine

Precaución: for Research Use Only

Formula: C₅H₇N₃
MW: 109.13 g/mol
MDL Number: MFCD01863677
CAS Number: 3435-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR