2-(Difluoromethoxy)-4-fluorobromobenzene 98%

Supplier: Ambeed
Warning

A110038-1G A110038-5G A110038-10G
76781-034EA 26 USD
76781-034 76781-036 77871-443
2-(Difluoromethoxy)-4-fluorobromobenzene 98%
2-(Difluoromethoxy)-4-fluorobromobenzene

Precaución: for Research Use Only

Formula: C₇H₄BrF₃O
MDL Number: MFCD11036070
CAS Number: 954235-83-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR