tert-Butyl-3-oxo-2,8-diazaspiro[4.5]decane-8-carboxylate 97%

Supplier: Ambeed
Warning

A109926-5G A109926-1G A109926-100MG A109926-250MG A109926-25G
76781-032EA 148 USD
76781-032 77570-556 77871-440 77871-441 77871-442
tert-Butyl-3-oxo-2,8-diazaspiro[4.5]decane-8-carboxylate 97%
tert-Butyl-3-oxo-2,8-diazaspiro[4.5]decane-8-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₂₂N₂O₃
MDL Number: MFCD11111226
CAS Number: 169206-67-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR