Spermine-(N,N'-bis(3-aminopropyl)-1,4-butanediamine) 90%
Supplier: Ambeed
|
Danger
|
Synonyms:
N1,N1'-(Butane-1,4-diyl)bis(propane-1,3-diamine), Spermine
A109610-5G
A109610-25G
A109610-1G
76781-020EA
122.01
USD
76781-020
77549-112
77570-546
Spermine-(N,N'-bis(3-aminopropyl)-1,4-butanediamine) 90%
Spermine-(N,N'-bis(3-aminopropyl)-1,4-butanediamine)
Precaución: for Research Use Only
Formula:
C₁₀H₂₆N₄ MW: 202.35 g/mol |
MDL Number:
MFCD00008215 CAS Number: 71-44-3 UN: 3259 ADR: 8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...