Tris[3,5-bis(trifluoromethyl)phenyl]phosphine 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Tris(3,5-bis(trifluoromethyl)phenyl)phosphine
A109462-100MG
A109462-1G
A109462-250MG
A109462-5G
A109462-25G
76781-012EA
15.92
USD
76781-012
76781-014
76781-016
76781-018
77666-594
Tris[3,5-bis(trifluoromethyl)phenyl]phosphine 95%
Tris[3,5-bis(trifluoromethyl)phenyl]phosphine
Precaución: for Research Use Only
Formula:
C₂₄H₉F₁₈P MW: 670.28 g/mol |
MDL Number:
MFCD00042428 CAS Number: 175136-62-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...