Ethyl-2-(5-bromobenzofuran-3-yl)acetate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 2-(5-bromobenzofuran-3-yl)acetate
A109237-100MG
A109237-1G
A109237-250MG
A109237-25G
A109237-5G
76780-998EA
95.49
USD
76780-998
76781-000
76781-002
76781-004
76781-006
Ethyl-2-(5-bromobenzofuran-3-yl)acetate 97%
Ethyl-2-(5-bromobenzofuran-3-yl)acetate
Precaución: for Research Use Only
Formula:
C₁₂H₁₁BrO₃ MW: 283.13 g/mol |
MDL Number:
MFCD09878709 CAS Number: 200204-85-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...