Ethyl-7-bromo-4-hydroxyquinoline-3-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 7-Bromo-4-hydroxy-3-quinolinecarboxylate

A109111-1G A109111-25G A109111-5G
76780-990EA 38.2 USD
76780-990 76780-992 76780-994
Ethyl-7-bromo-4-hydroxyquinoline-3-carboxylate 95%
Ethyl 7-bromo-4-hydroxyquinoline-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₀BrNO₃
MW: 296.1200 g/mol
MDL Number: MFCD00487280
CAS Number: 179943-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR