Tris[4-(trifluoromethyl)phenyl]phosphine 97%

Supplier: Ambeed
Warning

A108436-1G A108436-250MG A108436-5G A108436-25G
76780-960EA 25 USD
76780-960 76780-962 76780-964 77599-566
Tris[4-(trifluoromethyl)phenyl]phosphine 97%
Tris[4-(trifluoromethyl)phenyl]phosphine

Precaución: for Research Use Only

Formula: C₂₁H₁₂F₉P
MDL Number: MFCD00058883
CAS Number: 13406-29-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR