3-(Benzyl(isopropyl)amino)-1-(naphthalen-2-yl)propan-1-one hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A107403-10MG
A107403-1MG
A107403-50MG
A107403-5MG
76780-896EA
8
USD
76780-896
76780-898
76780-900
76780-902
3-(Benzyl(isopropyl)amino)-1-(naphthalen-2-yl)propan-1-one hydrochloride 97%
3-(Benzyl(isopropyl)amino)-1-(naphthalen-2-yl)propan-1-one hydrochloride
Precaución: for Research Use Only
Formula:
C₂₃H₂₆ClNO |
MDL Number:
MFCD04974534 CAS Number: 1021868-92-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...