Methyl-2,5-dibromothiazole-4-carboxylate 97%

Supplier: Ambeed
Warning

A107235-10G A107235-1G A107235-250MG A107235-5G
76780-888EA 156 USD
76780-888 76780-890 76780-892 76780-894
Methyl-2,5-dibromothiazole-4-carboxylate 97%
Methyl-2,5-dibromothiazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₅H₃Br₂NO₂S
MDL Number: MFCD08275712
CAS Number: 914347-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR