N-(tert-Butoxycarbonyl)-4-chloro-L-phenylalanine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Boc-Phe(4-Cl)-OH, N-Boc-4-chloro-L-phenylalanine
A107014-10G
A107014-250MG
A107014-25G
A107014-5G
A107014-1G
76780-872EA
81.69
USD
76780-872
76780-874
76780-876
76780-878
77666-558
N-(tert-Butoxycarbonyl)-4-chloro-L-phenylalanine 97%
N-(tert-Butoxycarbonyl)-4-chloro-L-phenylalanine
Precaución: for Research Use Only
Formula:
C₁₄H₁₈ClNO₄ MW: 299.75 g/mol |
MDL Number:
MFCD00057808 CAS Number: 68090-88-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...