Vanadyl(IV) acetylacetonate 98%

Supplier: Ambeed
Warning

Synonyms: Vanadium(IV) oxide acetylacetonate, Oxobis(2,4-pentanedionato-O,O')vanadium (IV), Vanadium(IV) bis(acetylacetonato)oxide, Vanadyl acetylacetonate, Di(2,4-pentandionato-O,O')vanadium(IV)oxid, Vanadium (IV) oxide bis(2,4-pentanedionate)

A105876-100G A105876-10G A105876-25G A105876-500G A105876-1000G
76780-804EA 23 USD
76780-804 76780-806 76780-808 76780-810 77666-552
Vanadyl(IV) acetylacetonate 98%
Vanadyl(IV) acetylacetonate

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₅V
MW: 265.16 g/mol
MDL Number: MFCD00000032
CAS Number: 3153-26-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR