(2-(2-Methyl-1H-imidazol-1-yl)phenyl)methanol 95%

Supplier: Ambeed
Warning

A105647-10G A105647-250MG A105647-5G
76780-796EA 229.16 USD
76780-796 76780-798 76780-800
(2-(2-Methyl-1H-imidazol-1-yl)phenyl)methanol 95%
(2-(2-Methyl-1H-imidazol-1-yl)phenyl)methanol

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂O
MW: 188.2300 g/mol
MDL Number: MFCD05864818
CAS Number: 914349-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR