Stevioside 95%

Supplier: Ambeed
Warning

A105391-10G A105391-25G A105391-25MG A105391-500G A105391-50MG A105391-5G A105391-100G
76780-784EA 12 USD
76780-784 76780-786 76780-788 76780-790 76780-792 76780-794 77565-842
Stevioside 95%
Stevioside

Precaución: For research use only.

Formula: C₃₈H₆₀O₁₈
MDL Number: MFCD00079561
CAS Number: 57817-89-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR