4-tert-Butyl-3,6-dichloropyridazine 97%

Supplier: Ambeed
Warning

A105125-1G A105125-250MG
76780-780EA 146.41 USD
76780-780 76780-782
4-tert-Butyl-3,6-dichloropyridazine 97%
4-tert-Butyl-3,6-dichloropyridazine

Precaución: for Research Use Only

Formula: C₈H₁₀Cl₂N₂
MW: 205.0800 g/mol
MDL Number: MFCD01938756
CAS Number: 22808-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR