(3-Fluoro-2-methylphenyl)acetic acid 98%

Supplier: Ambeed
Warning

A104970-10G A104970-250MG A104970-5G A104970-1G
76780-770EA 188 USD
76780-770 76780-772 76780-774 77666-536
(3-Fluoro-2-methylphenyl)acetic acid 98%
(3-Fluoro-2-methylphenyl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₂
MDL Number: MFCD03788545
CAS Number: 500912-16-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR