2,4,6-Trichloro-5-fluoropyrimidine 97%

Supplier: Ambeed
Warning

A104781-1G A104781-25G A104781-10G A104781-250MG
76780-764EA 33 USD
76780-764 76780-766 77666-530 77871-403
2,4,6-Trichloro-5-fluoropyrimidine 97%
2,4,6-Trichloro-5-fluoropyrimidine

Precaución: for Research Use Only

Formula: C₄Cl₃FN₂
MDL Number: MFCD11040435
CAS Number: 6693-08-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR