2-(trans-4-Aminocyclohexyl)propan-2-ol 97%
Supplier: Ambeed
|
Danger
|
A104744-100MG
A104744-10G
A104744-1G
A104744-250MG
A104744-5G
76780-754EA
43
USD
76780-754
76780-756
76780-758
76780-760
76780-762
2-(trans-4-Aminocyclohexyl)propan-2-ol 97%
2-(trans-4-Aminocyclohexyl)propan-2-ol
Precaución: for Research Use Only
Formula:
C₉H₁₉NO |
MDL Number:
MFCD11053512 CAS Number: 899806-45-2 UN: 3259 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...