tert-Butyl (4-bromothiazol-2-yl)(methyl)carbamate 95%

Supplier: Ambeed
Warning

A104541-250MG A104541-1G A104541-5G
76780-736EA 27 USD
76780-736 77666-514 77666-516
tert-Butyl (4-bromothiazol-2-yl)(methyl)carbamate 95%
tert-Butyl (4-bromothiazol-2-yl)(methyl)carbamate

Precaución: for Research Use Only

Formula: C₉H₁₃BrN₂O₂S
MDL Number: MFCD09878588
CAS Number: 1000576-79-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR