Tetrahydrofuran-3-ylmethanol 97%

Supplier: Ambeed
Warning

A104494-100G A104494-10G A104494-1G A104494-25G A104494-5G
76780-720EA 263 USD
76780-720 76780-722 76780-724 76780-726 76780-728
Tetrahydrofuran-3-ylmethanol 97%
Tetrahydrofuran-3-ylmethanol

Precaución: for Research Use Only

Formula: C₅H₁₀O₂
MDL Number: MFCD00075198
CAS Number: 15833-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR