N-(tert-Butoxycarbonyl)-4-bromo-D-phenylalanine 98%

Supplier: Ambeed
Warning

Synonyms: Boc-D-Phe(4-Br)-OH, N-Boc-4-bromo-D-phenylalanine

A104346-1G A104346-250MG A104346-25G A104346-5G A104346-100G A104346-10G
76780-702EA 7.43 USD
76780-702 76780-704 76780-706 76780-708 77599-524 77666-512
N-(tert-Butoxycarbonyl)-4-bromo-D-phenylalanine 98%
N-(tert-Butoxycarbonyl)-4-bromo-D-phenylalanine

Precaución: for Research Use Only

Formula: C₁₄H₁₈BrNO₄
MW: 344.21 g/mol
MDL Number: MFCD00270359
CAS Number: 79561-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR