2,3-Dihydrobenzo[b][1,4]dioxin-5-ol 97%

Supplier: Ambeed
Warning

A104058-10G A104058-1G A104058-25G A104058-250MG A104058-5G
76780-668EA 233.4 USD
76780-668 76780-670 76780-672 77666-500 77666-502
2,3-Dihydrobenzo[b][1,4]dioxin-5-ol 97%
2,3-Dihydrobenzo[b][1,4]dioxin-5-ol

Precaución: for Research Use Only

Formula: C₈H₈O₃
MW: 152.1500 g/mol
MDL Number: MFCD02103729
CAS Number: 10288-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR