2-Amino-3,5-dichloropyrazine 97%

Supplier: Ambeed
Warning

A103540-1G A103540-5G A103540-10G
76780-652EA 6.37 USD
76780-652 76780-654 77565-790
2-Amino-3,5-dichloropyrazine 97%
2-Amino-3,5-dichloropyrazine

Precaución: For research use only.

Formula: C₄H₃Cl₂N₃
MW: 163.9900 g/mol
MDL Number: MFCD11040179
CAS Number: 873-42-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR