5-Iodo-2-methyl-2H-indazole 98%

Supplier: Ambeed
Warning

A103433-100MG A103433-10G A103433-1G A103433-250MG A103433-5G
76780-642EA 47.75 USD
76780-642 76780-644 76780-646 76780-648 76780-650
5-Iodo-2-methyl-2H-indazole 98%
5-Iodo-2-methyl-2H-indazole

Precaución: for Research Use Only

Formula: C₈H₇IN₂
MW: 258.06 g/mol
MDL Number: MFCD11977518
CAS Number: 1150617-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR