2-Methoxypyrimidin-5-ol 98%

Supplier: Ambeed
Warning

A103337-100MG A103337-1G A103337-250MG
76780-626EA 66 USD
76780-626 76780-628 76780-630
2-Methoxypyrimidin-5-ol 98%
2-Methoxypyrimidin-5-ol

Precaución: for Research Use Only

Formula: C₅H₆N₂O₂
MDL Number: MFCD10696950
CAS Number: 91233-70-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR