3-Methoxy-4-nitrophenol 95%

Supplier: Ambeed
Warning

A103071-10G A103071-1G A103071-250MG A103071-25G A103071-5G
76780-598EA 204.76 USD
76780-598 76780-600 76780-602 76780-604 76780-606
3-Methoxy-4-nitrophenol 95%
3-Methoxy-4-nitrophenol

Precaución: for Research Use Only

Formula: C7H7NO4
MW: 169.1400 g/mol
MDL Number: MFCD11840329
CAS Number: 16292-95-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR