5-Aminobenzotriazole 98%

Supplier: Ambeed
Warning

Synonyms: 1H-Benzo[d][1,2,3]triazol-5-amine, 5-Amino-1H-benzotriazole

A103032-1G A103032-250MG A103032-5G A103032-25G A103032-100MG
76780-592EA 19 USD
76780-592 76780-594 76780-596 77666-492 77871-387
5-Aminobenzotriazole 98%
5-Aminobenzotriazole

Precaución: for Research Use Only

Formula: C₆H₆N₄
MDL Number: MFCD00047209
CAS Number: 3325-11-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR