2-Chloro-5-fluoropyridine-3-carbaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-5-fluoronicotinaldehyde

A103028-100MG A103028-10G A103028-1G A103028-250MG A103028-25G A103028-5G
76780-584EA 9 USD
76780-584 76780-586 76780-588 76780-590 77871-385 77871-386
2-Chloro-5-fluoropyridine-3-carbaldehyde 95%
2-Chloro-5-fluoropyridine-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₆H₃ClFNO
MDL Number: MFCD03095247
CAS Number: 851484-95-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR