1-(Pyrimidin-5-yl)ethanone 95%

Supplier: Ambeed
Warning

Synonyms: 5-Acetylpyrimidine

A102659-250MG A102659-5G A102659-1G
76780-566EA 26.53 USD
76780-566 77570-152 77592-086
1-(Pyrimidin-5-yl)ethanone 95%
1-(Pyrimidin-5-yl)ethanone

Precaución: for Research Use Only

Formula: C₆H₆N₂O
MW: 122.1246 g/mol
MDL Number: MFCD09038477
CAS Number: 10325-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR