N-(tert-Butoxycarbonyl)-3,4-difluoro-L-phenylalanine 97%

Supplier: Ambeed
Warning

A102246-1G A102246-250MG A102246-5G
76780-530EA 39 USD
76780-530 76780-532 76780-534
N-(tert-Butoxycarbonyl)-3,4-difluoro-L-phenylalanine 97%
N-(tert-Butoxycarbonyl)-3,4-difluoro-L-phenylalanine

Precaución: for Research Use Only

Formula: C₁₄H₁₇F₂NO₄
MDL Number: MFCD00672518
CAS Number: 198474-90-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR