Boc-(R)-3-amino-4-(3,4-dichlorophenyl)butyric acid 98%

Supplier: Ambeed
Warning

Synonyms: (R)-3-((tert-Butoxycarbonyl)amino)-4-(3,4-dichlorophenyl)butanoic acid

A101822-100MG A101822-1G A101822-250MG
76780-520EA 44.56 USD
76780-520 76780-522 76780-524
Boc-(R)-3-amino-4-(3,4-dichlorophenyl)butyric acid 98%
Boc-(R)-3-amino-4-(3,4-dichlorophenyl)butyric acid

Precaución: for Research Use Only

Formula: C₁₅H₁₉Cl₂NO₄
MW: 348.23 g/mol
MDL Number: MFCD01860949
CAS Number: 269396-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR