Z-Pro-OSu 95%

Supplier: Ambeed
Warning

Synonyms: 1-Benzyl 2-(2,5-dioxopyrrolidin-1-yl) (S)-pyrrolidine-1,2-dicarboxylate

A101400-100G A101400-25G A101400-5G
76780-492EA 308.73 USD
76780-492 76780-494 76780-496
Z-Pro-OSu 95%
Z-Pro-OSu

Precaución: for Research Use Only

Formula: C₁₇H₁₈N₂O₆
MW: 346.339 g/mol
MDL Number: MFCD00042757
CAS Number: 3397-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR