Z-Hyp-OMe 97%

Supplier: Ambeed
Warning

Synonyms: N-Cbz-4-trans-hydroxy-L-proline methyll ester, N-Benzyloxycarbonyl-4-trans-hydroxy-L-proline methyll ester, Z-Hyp-OMe, N-Carbobenzoxy-4-trans-hydroxy-L-proline methyll ester, (2S,4R)-1-Benzyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate, N-Benzyloxycarbonyl-4-trans-hydroxy-L-proline methyl ester

A101081-100G A101081-10G A101081-25G A101081-500G A101081-5G
76780-454EA 58 USD
76780-454 76780-456 76780-458 76780-460 76780-462
Z-Hyp-OMe 97%
Z-Hyp-OMe

Precaución: for Research Use Only

Formula: C₁₄H₁₇NO₅
MDL Number: MFCD00055851
CAS Number: 64187-48-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR