SB216763 98%

Supplier: Ambeed
Warning

Synonyms: 3-(2,4-Dichlorophenyl)-4-(1-methyl-1H-indol-3-yl)-1H-pyrrole-2,5-dione, SB-216763

A100785-10MG A100785-1MG A100785-5MG A100785-100MG A100785-1G
76780-398EA 57.29 USD
76780-398 76780-400 76780-402 77666-458 77666-460
SB216763 98%
SB216763

Precaución: for Research Use Only

Formula: C₁₉H₁₂Cl₂N₂O₂
MW: 371.2 g/mol
MDL Number: MFCD09753369
CAS Number: 280744-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR