4-Fluoro-3-(trifluoromethyl)benzyl bromide 97%

Supplier: Ambeed
Danger

Synonyms: 4-(Bromomethyl)-1-fluoro-2-(trifluoromethyl)benzene, 1-(Bromomethyl)-4-fluoro-3-(trifluoromethyl)benzene

A998987-1G A998987-25G A998987-5G
76777-088EA 6 USD
76777-088 76777-090 76777-092
4-Fluoro-3-(trifluoromethyl)benzyl bromide 97%
4-Fluoro-3-(trifluoromethyl)benzyl bromide

Precaución: for Research Use Only

Formula: C₈H₅BrF₄
MDL Number: MFCD00061177
CAS Number: 184970-26-1
UN: 1760
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR