1-Acetyl-5-bromo-7-nitroindoline 98%

Supplier: Ambeed
Warning

Synonyms: 1-(5-Bromo-7-nitroindolin-1-yl)ethanone

A984034-1G A984034-25G
76777-070EA 13.8 USD
76777-070 76777-072
1-Acetyl-5-bromo-7-nitroindoline 98%
1-Acetyl-5-bromo-7-nitroindoline

Precaución: for Research Use Only

Formula: C₁₀H₉BrN₂O₃
MW: 285.1 g/mol
MDL Number: MFCD00056018
CAS Number: 62368-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR