5-Amino-1,3-dimethyl-1H-benzo[d]imidazol-2(3H)-one 95%

Supplier: Ambeed
Warning

A980494-100MG A980494-1G A980494-250MG
76777-064EA 31 USD
76777-064 76777-066 76777-068
5-Amino-1,3-dimethyl-1H-benzo[d]imidazol-2(3H)-one 95%
5-Amino-1,3-dimethyl-1H-benzo[d]imidazol-2(3H)-one

Precaución: for Research Use Only

Formula: C₉H₁₁N₃O
MW: 177.20 g/mol
MDL Number: MFCD00462960
CAS Number: 53439-88-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR